Anti-Ly-6A/E Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: D7]

Supplier: BIOGEMS INTERNATIONAL INC.
82912-77-50
75843-610EA 668.69 CAD
75843-610
Anti-Ly-6A/E Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: D7]
Antibodies
The D7 monoclonal antibody specifically reacts with mouse Ly-6A/E (Sca-1), a 18 kDa GPI-linked surface protein. It is expressed on multipotent hematopoietic stem cells (HSC) and subsets of B and T cells and involved in B and T cell lymphocyte responses. Splenocytes from Ly-6.2 strains exhibit a larger number of D7-positive cells than those from the Ly-6.1 strains.

Type: Primary
Antigen: Ly-6A/E
Clonality: Monoclonal
Clone: D7
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR