Drying Chambers, Labconco®
Supplier: Labconco
7522800 7522900 7318700 7318702 7318800 7318802 7522801 7522901
26672-140EA 4613.6 CAD
26672-140 26672-152 30181-309 30181-311 30181-401 30181-403 82010-512 82010-514 75841-008 75841-010 75841-012 75841-014 75841-034 75841-036
Drying Chambers, Labconco®
Desiccators Desiccators, Glass
Designed for use with FreeZone® Freeze Dryers. Provide strength, durability and corrosion resistance.
  • Chambers are available in regular, heated, and clear models
The 12- and 16-port drying chambers are made of Type 304 stainless steel with a clear acrylic top.
Supplied with neoprene valves which accommodate both 1.3 cm (1/2") and 1.9 cm (3/4") adapters. Lids have a neoprene gasket around the edge to assure a leakproof vacuum seal. Models with PTFE-coated interior provide corrosion resistance. The 12.0" diameter clear acrylic chambers (75841-010 andMore Product Information