PRO Internal Baths with Command Touch, Lauda-Brinkmann
Supplier: Lauda-Brinkmann
L000317 L000318 L000454 L000550 L000551 L000552
75840-820EA 34097.33 CAD
75840-820 75840-822 75840-824 75840-826 75840-828 75840-840 75840-842 75840-856 75840-860 75840-862 75840-864
PRO Internal Baths with Command Touch, Lauda-Brinkmann
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
Our PRO units have advantages such as low filling volume combined with powerful Varioflex pump, resulting in rapid temperature change. Environment Friendly (GWP).
  • Large color touch screen (5.7")
  • Programmer with 100 programs; each with 50 segments
  • Weekly timer and count-down function
  • Data logging; export to USB stick
  • Individual user rights up to 20 users
  • Menu navigation in 8 languages
  • Removable touch command
  • Temperature in °C or °F
LAUDA PRO Thermostats are optimized for interal bath applications and offer excellent temperature stability
The COMMANDMore Product Information