Rotary Pulse Encoders, Nidec Shimpo
Supplier: Nidec Shimpo America
RE1B-60F RE1-BRKT RE2B-30F RE1B-1000F RE1B-600F RE2B-600F BRACKET-M18
75840-646EA 554.98 CAD
75840-646 75840-648 75840-650 75840-668 75840-670 75840-672 75840-688
Rotary Pulse Encoders, Nidec Shimpo
Data Loggers
Encoders are ideal for use with panel mount tachometers and counters to monitor motor operation.
  • Rotary encoders available from 60 to 1000 ppr, with 5000 rpm maximum capability
  • Quadrature versions available
  • RE series encoders are ideal for usage with SHIMPO’s panel mount tachometers and counters
  • Base or face mounting
  • ABS housing enclosure
RE1B-60F: 60 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-600F: 600 digital output pulses per shaft revolution, maximum speed of 5000 rpm.
RE1B-1000F: 1000 digital output pulses per shaftMore Product Information