Proximity Sensors, Nidec Shimpo
Supplier: Nidec Shimpo America
BI2-S12 MCS-3109 SE-G2
75840-644EA 220.58 CAD
75840-644 75840-658 75840-674 75840-696
Proximity Sensors, Nidec Shimpo
Data Loggers
Ideal for gear speed measurement.
  • Rugged Proximity Sensors to measure position, movement, or rotational speeds
  • Economical solution for detecting metal in front of a sensing surface
  • Metal objects adjacent to or behind will not normally be detected
Model DJ2 features a pre-wired 17' cable assembly, zero-sensing capability, and small package size
Model B12-S12 is a three-wire DC normally open NPN (sinking) type proximity switch which features a short circuit and overload protection plus LED output indicator.
Model SE-G is a solid state proximityMore Product Information