Manual Lever and Wheel Operated Force Test Stands, SEALS USA INC
Supplier: Nidec Shimpo America
FGS-50S FGS-100L
75840-592EA 1715.58 CAD
75840-592 75840-594 75840-596
Manual Lever and Wheel Operated Force Test Stands, SEALS USA INC
Dynamometers
Hand wheel and lever operated Force Test Stands feature adjustable clearance settings that enable testing on a wide range of samples. The compact design reduces footprint.
  • Versatile bottom mounting plate permits acceptance of both standard and custom-made attachments
  • Adjustable clearance setting enables testing on a wide range of samples
  • Precision gear movement provides a consistent linear motion that produces repetitively accurate results
  • Modular, compact design reduces footprint, weight, and cost
  • Sturdy all steel construction provides exceptional
More Product Information