Securtainer™ Specimen Containers, Polypropylene, Simport Scientific
Supplier: Simport Scientific
C576-20MA C576-40MA C576-60MA C576-120MA C575-120MA C575-20MA C575-40MA C575-60MA C575-90MA C575-20MA C575-40MA C575-60MA C575-90MA C577-20W C577-40W C577-60W C577-90W C577-120W
89213-156CS 427.33 CAD
89213-156 89213-158 89213-160 89213-164 CA11027-884 CA11027-886 CA11027-888 CA11027-890 CA11027-892 89500-336 89500-338 89500-340 89500-342 75840-150 75840-152 75840-154 75840-156 75857-052
Securtainer™ Specimen Containers, Polypropylene, Simport Scientific
Sample Containers General Purpose Containers
Securtainer™ Specimen Containers are designed for collection, transport and storage of specimens
Available in three unique models, containers and caps are manufactured without the use of plasticisers or mold release agents. Material used in manufacturing is free from latex. All containers are 95kpa compliant. Available in many sizes from 20 to 120mL. Non-sterile.
The SecurTainer™ I sample container incorporates a unique push tab tamper evident locking mechanism. Once fully screwed, the tamper evident ring will detach itself from the cap showing the container was opened. The Securtainer™More Product Information