VWR® Advanced Dry Block Heaters
Supplier: VWR International
75838-270 75838-278 75838-280 75838-282 75838-284 75838-294 75838-296 75838-306 75838-308 75838-316
75838-270EA 1721.18 CAD
75838-270 75838-278 75838-280 75838-282 75838-284 75838-294 75838-296 75838-306 75838-308 75838-316
VWR® Advanced Dry Block Heaters
Block Heaters Digital Block Heaters
Designed for applications that require repeatable results and superior temperature stability.
  • New design with smaller footprint
  • Exceptional temperature uniformity and stability
  • Heated lid model reduces condensation in sample lids
  • LED display for temperature and time
  • Heats to 120 °C (245 °F)
  • Holds interchangeable, modular heating blocks
  • Five-year warranty on parts and labor
  • Available with a NIST Traceable, ISO/IEC 17025 acredited calibration certificate from Troemner
  • Optional temperature probe kit
Multi-purpose units are ideal for incubation andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing Waste Reducing Waste Reducing Waste Reducing Waste Reducing
Temperature range Ambient +5 to 120 °C (248 °F) Ambient +5 to 100 °C (212 °F) Heated Lid Model Ambient +5 to 120 °C (248 °F) Ambient +5 to 120 °C (248 °F) Ambient +5 to 120 °C (248 °F)
Temperature stability ±0.1°C ±0.1°C ±0.1°C ±0.1°C ±0.1°C
Temperature uniformity ±0.1 °C (across similar blocks at 37 °C) ±0.1 °C (across similar blocks at 37 °C) ±0.2 °C (across similar blocks at 37 °C) ±0.3 °C (across similar blocks at 37 °C)
Environmental certification RoHS RoHS RoHS RoHS RoHS