FlexTech™ Lightweight Y9289 Synthetic Knit Gloves with Double NBR Coating, Wells Lamont
Supplier: Wells Lamont
Y9289XS Y9289S Y9289M Y9289L Y9289XL Y9289XXL
75836-500PK 210.2 CAD
75836-500 75836-502 75836-540 75836-542 75836-544 75836-546
FlexTech™ Lightweight Y9289 Synthetic Knit Gloves with Double NBR Coating, Wells Lamont
Gloves
FlexTech™ Y9289 synthetic knit gloves are double coated for excellent gripping.
  • Light weight synthetic kni
  • Double full NBR coating
  • Excellent grip
  • Excellent for general assembly
FlexTech™ Y9289 synthetic knit gloves are light weight. They feature a full double NBR coating for an extra layer of protection against liquids. The sandy nitrile coated palm and fingertips affords excellent gripping with great flexibility and dexterity. These gloves are excellent for general assembly, automotive, painting, manufacturing and the handling of small, wet and oilyMore Product Information