Mini-300 Orbital Genie™, Scientific Industries
Supplier: Scientific Industries
SI-M1500 SI-M1600 SI-M1700
75836-244EA 2998.26 CAD
75836-244 75836-252 75836-260
Mini-300 Orbital Genie™, Scientific Industries
Shakers Orbital Shakers
Powerful high speed orbital shakers for use with virtually any size or shape container in three different configurations.
  • Super compact design
  • Powerful motor for shaking speed up to 300 rpm
  • Three models provides choice
  • Adhering mat model is ideal for open containers, bottles, flasks, or beakers
  • Ratcheting clamps model provides flexibility and stronghold of various size and shape containers
  • Standard flask clamps model can be used with clamps sold separately
Heavy duty, low profile, compact footprint Orbital Shakers available in 3 differentMore Product Information