8095 Calibration Mix B (7 Components), Restek
Supplier: RESTEK CORP
31608
75835-704EA 107.36 CAD
75835-704
8095 Calibration Mix B (7 Components), Restek
Standards Organic Standards
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable