Chrysene, Restek

Supplier: RESTEK CORP

31275
75835-640EA 57.67 CAD
75835-640
Chrysene, Restek
Standards Organic Standards

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Order Now

SPECIFICATIONS

Description Chrysene
Environmentally Preferable
CAS No. 218-01-9
Formula C₁₈H₁₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR