QuEChERS Single-Component Reference Standards, Restek

Supplier: RESTEK CORP

33257 33258 33259
75835-194EA 52.09 CAD
75835-194 75835-196 75835-198 75835-200 75835-204 75835-206
QuEChERS Single-Component Reference Standards, Restek
Standards Organic Standards

  • Ready to use for QuEChERS extractions—no dilutions necessary
  • Support for GC and LC with MS, MS/MS, and selective detectors

Order Now

SPECIFICATIONS

Description PCB 28 PCB 52 Triphenylphosphate Tris(1,3-dichloroisopropyl)phosphate
Environmentally Preferable
CAS No. 7012-37-5 35693-99-3 115-86-6 13674-87-8
Formula C₁₂H₇Cl₃ C₁₂H₆Cl₄ C₁₈H₁₅O₄P C₉H₁₅Cl₆O₄P

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR