Chlorinated Acids by LC, Mix B (8 Components), Restek
Supplier: RESTEK CORP
32430
75834-984EA 169.91 CAD
75834-984
Chlorinated Acids by LC, Mix B (8 Components), Restek
Standards Organic Standards
Consists of eight components, those are 2,4-db, 3,5-dichlorobenzoic acid, dinoseb, MCPA, MCPP, 4-nitrophenol, pentachlorophenol and 2,4,5-t.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.