DRO/ORO Calibration Standard, Restek
Supplier: RESTEK CORP
31831
75834-800EA 82.16 CAD
75834-800
DRO/ORO Calibration Standard, Restek
Standards Petroleum Industry Standards
This consists of 10W30 motor oil:diesel fuel #2 (1:1 blend).