CELLSPIN Spinner System, Argos Technologies
Supplier: Antyila Scientific
13051-00 24406-17 24406-18 13051-58 13051-60 13051-62 13051-64 24406-19 24406-20 24406-21 24406-24 24406-25
75834-420EA 15767.89 CAD
75834-420 75834-422 75834-424 75834-426 75834-428 75834-430 75834-432 75834-434 75834-436 75834-438 75834-440 75834-442
CELLSPIN Spinner System, Argos Technologies
Shakers Rotating/Rocking Shakers
CELLSPIN a reliable cell culture system.
  • Culture volumes from 100 to 1000 ml
  • Can be individually programmed
  • Equipped with an alarm system
The CELLSPIN is a stirrer for cultivating cells in spinner bottles with culture volumes from 100 to 1000 ml. The speed (5-75 rpm) and other stirring parameters can be individually programmed and modified at any time via the control unit, which is located outside of the incubator. In case of power failure or accidental disconnection of control and spinner units an alarm system is activated. The corrosion-free andMore Product Information