4,4'-DDD, Restek

Supplier: RESTEK CORP

32201
75834-230EA 56.79 CAD
75834-230
4,4'-DDD, Restek
Standards Organic Standards

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.

Order Now

SPECIFICATIONS

Description 4,4'-DDD
Environmentally Preferable
CAS No. 72-54-8
Formula C₁₄H₁₀Cl₄

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR