XPR Microbalances, METTLER TOLEDO®
Supplier: Mettler Toledo
30279285 30279296 30279267 30279196 30279276
75832-778EA 63827.5 CAD
75832-778 75832-780 75832-782 75832-842 75832-844
XPR Microbalances, METTLER TOLEDO®
Balances and Scales
METTLER TOLEDO’s new XPR microbalances incorporate a range of innovative new technologies to deliver the highest accuracy in their class and help make your micro-weighing tasks simpler, safer and more secure.
  • Capacities from 2.1 g to 10.1 g
  • Readabilities from 0.1µg
  • Glass draft shield with motorized drive for precise weighing even in unstable environments.
  • Compact design with small footprint.
  • Fully automatic adjustment using internal weights.
  • Built-in level sensor and leveling aid for fast and easy leveling.
  • Capacitive color TFT touch screen
  • Two-terminal
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Communication interface RS-232, USB Host, USB Device
Voltage 12 VDC ±3%; 2.25 A, max. ripple: 80 mVpp