C-Flex® Tubing, Formulation 082, Saint-Gobain
Supplier: Saint Gobain Performance Plastics
082-125-2 082-375-2 082-500-3 082-125-2S 082-188-2S 082-250-3S 082-250-4S 082-313-3S 082-188-2M 082-250-4M 082-375-2M 082-375-3M 082-500-4M 082-250-4
75830-474PK 181.02 CAD
75830-474 75831-176 75831-196 75831-206 75831-208 75831-210 75831-212 75831-218 75831-220 75831-222 75831-224 75831-226 75831-228 75831-230 75831-232 75831-234 75831-236 75831-238 75831-240 75831-242 75831-244 75831-246 75831-248 75831-250 75831-252 75831-254 75831-256 75831-356 75831-358
C-Flex® Tubing, Formulation 082, Saint-Gobain
Tubing Transfer Tubing
C-Flex® is a unique, patented thermoplastic elastomer specifically designed to meet the critical demands of the pharmaceutical, research, biopharmaceutical and diagnostics industries. C-Flex® biopharmaceutical tubing has been used by many of the world’s leading biotech and pharmaceutical processing companies.
  • Non-pyrogenic, non-cytotoxic, non-hemolytic
  • Chemically resistant to concentrated acids and alkalis
  • Low platelet adhesion, low protein binding
  • Ultra-smooth inner bore
  • Available in clear or opaque formulations
  • Lot traceable, safe disposal through
More Product Information