Urine Drug Screening, Hydrocodone Assay, MedTest DX
Supplier: MEDTEST DX, INC.
10018054 10018053
75830-334EA 5414.6 CAD
75830-334 75831-162
Urine Drug Screening, Hydrocodone Assay, MedTest DX
Drug Tests
The Hydrocodone assay is intended for the qualitative and semi-quantitative detection and estimation of hydrocodone and its metabolites in human urine at a cutoff of 300 ng/ml.
  • Supplied as a liquid ready-to-use homogeneous enzyme immunoassay
The semi-quantitative mode enables to determine an appropriate dilution of specimen for confirmation by a confirmatory method such as LC-MS/MS or GC-MS and permitting laboratories to establish quality control measures. This assay provides a preliminary analytical test result. A more specific alternative chemicalMore Product Information