Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Life Sciences
PFR062X125PPAF PFR062X250PPAF PFC125PPAF PFT125PPAF PFMS125PPAF PFR125X187PPAF PFR125X250PPAF PFX125PPAF PFC187PPAF PFY250PPAF PFT250PPAF PFX250PPAF PFE250PPAF PF135250PPAF PFC312PPAF PFY312PPAF PFT312PPAF PFY375PPAF PFT375PPAF PFX375PPAF PFE375PPAF PFLS375PPAF PFMS375PPAF PFC375PPAF PFR375X500PPAF PF135375PPAF PFY500PPAF PFT500PPAF PFX500PPAF PFE500PPAF PFLS500PPAF PFMS500PPAF PFC500PPAF PFMS125PVDF PFC125PVDF PFT125PVDF PFX125PVDF PFR125X187PVDF PFR125X250PVDF PFX187PVDF PFT187PVDF PFC187PVDF PFR187X312PVDF PFR187X375PVDF PFY250PVDF PFT250PVDF PFX250PVDF PFE250PVDF PFLS250PVDF PFMS250PVDF PFC250PVDF PFR250X375PVDF PF135250PVDF PFY312PVDF PFT312PVDF PFC312PVDF PFY375PVDF PFT375PVDF PFX375PVDF PFE375PVDF PFLS375PVDF PFMS375PVDF PFC375PVDF PFR375X500PVDF PF135375PVDF PFT135062 PFT135125 PFT135187 PFT135312 PFT187PPAF PFX187PPAF PFR187X312PPAF PFR187X375PPAF PFLS250PPAF PFMS250PPAF PFC250PPAF PFR250X375PPAF PFR250X500PPAF PF135500PPAF PFMS625PPAF PFLS750PPAF PFT750PPAF PFR062X125PVDF PFR062X250PVDF PFY125PVDF PFY500PVDF PFT500PVDF PFX500PVDF PFE500PVDF PFC500PVDF PFR250X500PVDF PF135500PVDF PFMS625PVDF PFT750PVDF
75830-870BG 395.32 CAD
75830-870 75830-872 75830-874 75830-876 75830-878 75830-880 75830-882 75830-884 75830-886 75830-888 75830-890 75830-892 75830-894 75830-896 75830-898 75830-900 75830-902 75830-904 75830-906 75830-908 75830-910 75830-912 75830-914 75830-916 75830-918 75830-920 75830-922 75830-924 75830-926 75830-928 75830-962 75830-964 75830-966 75830-968 75830-970 75830-972 75830-974 75830-976 75830-978 75830-980 75830-982 75830-984 75830-986 75830-988 75830-990 75830-992 75830-994 75830-996 75830-998 75831-000 75831-002 75831-004 75831-006 75831-008 75831-010 75831-012 75831-014 75831-016 75831-018 75831-020 75831-022 75831-024 75831-026 75831-028 75831-030 75831-032 75831-034 75831-036 75831-038 75831-040 75831-042 75831-044 75831-046 75831-048 75831-066 75831-068 75831-070 75831-072 75831-074 75831-076 75831-078 75831-080 75831-082 75831-084 75831-086 75831-088 75831-090 75831-092 75831-094 75831-096 75831-098 75831-100 75831-102 75831-104 75831-106 75831-108 75831-110 75831-112 75831-114 75831-116 75831-118 75831-120
Pure-Fit® SIB® Smooth Inner Bore Systems, Saint-Gobain Performance Plastics
Fittings and Connectors
Pure-Fit SIB is a smooth inner bore (SIB) hose-barb connector that provides a seamless transition between tubing and fittings throughout the fluid path.
  • Eliminates entrapment and leak points that occur with traditional fitting systems
  • Complete 360° compression along the entire fitting system
  • No excessive voids at the interface or in the fluid path
  • Utilizes the patented BarbLock retention and seal system
  • Compatible with Sani-Tech, C-Flex and PharMed flexible tubing
Available in both animal-free polypropylene (PP) material, which is gamma stable andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable