Avantor® Alltima, Phenyl HPLC Columns
Supplier: Avantor
81188 88087 88088 88092 88093 96442/N 96082/N AL-5PH-300DE
75797-282EA 1815.06 CAD
75797-282 75797-284 75797-286 75797-288 75797-290 75797-294 75797-298 75797-300 75797-302 75797-304 75830-372 75830-850 76398-446
Avantor® Alltima, Phenyl HPLC Columns
Chromatography Columns
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L11
Less hydrophobic than C18 phase. Selective for aromatic compounds.
Order Now
SPECIFICATIONS
Description 75797-304
Environmentally Preferable
Carbon load 7.5%
Endcapped Yes
Surface area 340 m2/g
USP code L11