Sapphire PCR Tubes and Strips, Polypropylene, Greiner Bio-One
Supplier: Greiner Bio-One
373270 373273 671201 671221 671231 671241 673210 673271 673273 673274 673275 673276 673277 673280 673281 673282 673283 673284 673285 682201 682273 682274 682275 682276 682277 682281 683201 683271 683273 683274 683275 683276 683277 684201 373281 373275 373276 373277 671274 671281 373274 671275 671276 673286 673287 373292 671277 671273 373250 373293 683281 608281
89204-786CS 472.32 CAD
89204-786 10859-842 75796-108 75796-118 75796-120 75796-122 75796-124 75796-126 75796-128 75796-130 75796-132 75796-134 75796-136 75796-138 75796-140 75796-142 75796-144 75796-146 75796-148 75796-150 75796-152 75796-154 75796-156 75796-158 75796-160 75796-162 75796-164 75796-166 75796-168 75796-170 75796-172 75796-174 75796-176 75796-178 75816-118 75816-120 75816-122 75816-124 75816-126 75816-128 75820-360 75830-540 75830-542 75830-552 75830-554 75830-556 75830-558 75830-574 75830-576 75831-386 75831-388 75786-257
Sapphire PCR Tubes and Strips, Polypropylene, Greiner Bio-One
Tubes PCR Tubes
Sapphire PCR Tubes and strips feature attached or unattached caps in either domed or flat designs, and frosted caps are available for easy labeling.
  • Manufactured out of medical-grade polypropylene without interfering additives
  • Thin-walled for efficient heat transfer during amplification
  • Options for real-time/quantitative PCR
  • Comprehensive thermal cycler compatibility
  • Packaging labeled with product symbols for easier working and warehousing
Maximum volume capacity ranges between models. Tube strips are available in formats of standard or low profile.
More Product Information