BioFit Static Control ESD Chairs and Stools
Supplier: BioFit
AEA-M-RK-K-EBO BTS-H-HG-KNAVY BTS-H-R-K-KBLK BTS-L-R-K-NAVY BTS-M-HG-K-KBK BTSMHTXXKCNAV EEA-L-R-K-MBLU EEA-L-R-K-NAVY 75821-108 75821-122 75821-144 75821-174 75821-196 75821-200 75821-204 75821-242 75821-280 75821-302 75821-306 75821-364 75821-368 75821-404 75821-410 75821-414 75821-428 75821-446 75821-508
75796-458EA 2745.11 CAD
75796-458 75796-722 75796-732 75796-764 75796-788 75796-792 75796-872 75796-874 75821-108 75821-122 75821-144 75821-174 75821-196 75821-200 75821-204 75821-242 75821-280 75821-302 75821-306 75821-364 75821-368 75821-404 75821-410 75821-414 75821-428 75821-446 75821-508
BioFit Static Control ESD Chairs and Stools
Chairs
BioFit’s ESD chairs are made to meet critical static control environments and still provide user comfort, health and wellbeing.
  • Chrome finish
  • Spring-loaded contact ball bearing assembly assures continuity thoughout the chair
  • Drag wire for added continuity from the chair to the ground
  • Ergonomic comfort and support
  • Chairs feature height and tilt adjustable backrests with lumbar support
  • Backrest has electrically conductive 0.125" polypropylene rear panel
  • Dual-wheel resistance ESD casters enhance user safety by inhibiting seating from rolling away on hard
More Product Information