Gordon Brush® Metal Free Tube Brushes
Supplier: Gordon Brush
710212 710312 710512 710612 710712 710812 710518 710618 710718 710818 711018 711218 710224 710324 711024 711224 720712 720812 722012 722512 720718 720818 721018 721218 721524 721824 722024 722524 730712 731012 731212 731512 731718 732018 731224 731724 732024 732524 733024 740524 740724 741024 740536 740736 741036 740548 740748 741048 721518 721818 722018 722518 720724 720824 721024 721224 731712 732012 732512 733012 730718 731018 731218 731518 711012 711212 710218 710318 710524 710624 710724 710824 721012 721212 721512 721812 732518 733018 730724 731024
75816-502PK 435.92 CAD
75816-502 75816-504 75816-506 75816-508 75816-510 75816-512 75816-514 75816-516 75816-518 75816-520 75816-522 75816-524 75816-526 75816-528 75816-530 75816-532 75816-534 75816-536 75816-538 75816-540 75816-542 75816-544 75816-546 75816-548 75816-550 75816-552 75816-554 75816-556 75816-558 75816-560 75816-562 75816-564 75816-566 75816-568 75816-570 75816-572 75816-574 75816-576 75816-578 75816-580 75816-582 75816-584 75816-586 75816-588 75816-590 75816-592 75816-594 75816-596 75816-598 75816-600 75816-602 75816-604 75816-606 75816-608 75816-610 75816-612 75816-614 75816-616 75816-618 75816-620 75816-622 75816-624 75816-626 75816-628 75816-630 75816-636 75816-638 75816-640 75816-642 75816-644 75816-646 75816-648 75816-650 75816-652 75816-654 75816-656 75816-658 75816-660 75816-662 75816-664 75816-666
Gordon Brush® Metal Free Tube Brushes
Brushes
Metal Free Tube Brushes are manufactured with 100% food grade polypropylene handle and bristles. Polypropylene is resistant to acids, alkalis, alcohols, and many petroleum distillates. These brushes can be used to clean the inside of straight or curved glassware, tubes, pipes, valves, and fittings. They are ideally suited for moderate to aggressive scrubbing. Brushes will not scratch most surfaces, and the fused bristles help minimize the growth of bacteria. Available in white only.
  • 100% food grade polypropylene handle and bristles
  • Designs available to
More Product Information