Vydac 208TP C8, HPLC Columns
Supplier: Avantor
208TP5210 208TP5215 208TP5415 208TP52 208TP54 208TP510 208TP1010 208TP1022 208GCC1210 208FSK510 208FSK1210 208TP101510 208TP152022 208GD104/N 208GD34/N 208GD52/N 208GD54/N 208GK34/N 208GK52/N 208GK54/N
75798-296EA 1584.04 CAD
75798-296 75798-298 75798-300 75798-302 75798-316 75798-318 75798-320 75798-322 75798-324 75798-326 75798-328 75798-330 75798-332 75798-334 75798-336 75798-338 75798-340 75798-342 75798-348 75798-350 75798-352 75798-354 75812-924 75812-926 75812-928 75812-930 75813-000 75813-002 75813-004 75813-006 75813-008 75813-010 75813-012 75830-644 75830-646 75830-648 75831-180 75831-184 75831-186 75831-188 75831-192 76398-032 76398-034 76398-036 76398-040 76398-044
Vydac 208TP C8, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L7
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 to 20K MW.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of theMore Product Information