Sodium phosphate, anhydrous ≥97.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: tri-Sodium phosphate, Sodium phosphate tribasic, Trisodium phosphate

S1276-45KGBL
75812-220EA 4834.48 CAD
75812-220
Sodium phosphate, anhydrous ≥97.0% FCC
tri-Sodium phosphate

Sodium Phosphate, Tribasic, Anhydrous, FCC is used as an acidity regulator (buffering agent), emulsifier, thickening agent, nutrition enlargement agent and sequestrant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: Na₃O₄P
Storage Temperature: Ambient
MDL Number: MFCD00003510
CAS Number: 7601-54-9
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay (Na₃PO₄; Ignited Basis) 97.0 %
Arsenic max.3 mg/kg
Fluoride max. 0.005 %
Insoluble Substances max. 0.2 %
Lead max. 4 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR