di-Potassium hydrogen phosphate, anhydrous ≥98.0% FCC

Supplier: Spectrum Chemical Mfg. Corp.

PO205-12KG PO205-2.5KG PO205-45KGBL PO205-500GM
75812-074EA 2274.88 CAD
75812-074 75812-076 75812-078 75812-080
di-Potassium hydrogen phosphate, anhydrous ≥98.0% FCC
di-Potassium hydrogen phosphate

Potassium Phosphate, Dibasic, Anhydrous, FCC is used as an acidity regulator, antioxidant, sequestrant and stabilizer in food and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: K₂HPO₄
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (K₂HPO₄) 98.0 % min.
Arsenic (As) max.3 mg/kg
Fluoride max. 10 mg/kg
Insoluble Substances max. 0.2 %
Lead (Pb) max. 2 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR