Bole white (kaolin), powder FCC

Supplier: Spectrum Chemical Mfg. Corp.

K1004-12KG K1004-50KGBL
75811-446EA 2754.65 CAD
75811-446 75811-448
Bole white (kaolin), powder FCC
Bole white (Kaolin)

Kaolin, Powder, FCC is used as a whitener in toothpaste. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: Al₂SiO₇·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7

Order Now

Specification Test Results

Acid-Soluble Substances max. 2.0 %
Arsenic (As) max.3 mg/kg
Carbonate (CO₃) To Pass Test
Iron (Fe) To Pass Test
Lead (Pb) max. 10 mg/kg

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR