Eugenol USP

Supplier: Spectrum Chemical Mfg. Corp.

EU110-20LTBL
75811-108EA 3611.09 CAD
75811-108
Eugenol USP
Eugenol

EUGENOL, USP, is used in perfumeries, flavorings, essential oils and in medicine as a local antiseptic and anesthetic. Eugenol can be combined with zinc oxide to form a material – known as zinc oxide eugenol – which has restorative and prosthodontic applications in dentistry. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities. Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₀H₁₂O₂
Density: 1.066
Storage Temperature: Ambient
MDL Number: MFCD00008654
CAS Number: 97-53-0

Order Now

Specification Test Results

Solubility in 70% Alcohol To Pass Test
Specific Gravity @ 25°C 1.064 - 1.070
Distilling Range 250 - 255 °C
Refractive Index @ 20°C 1.540 - 1.542
Heavy Metals (As Pb) max. 0.004 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR