Venusil AQ C18 HPLC Columns, Bonna-Agela Technologies
Supplier: Phenomenex
VA930105-0 VA930502-0 VA931003-0 VA931502-0 VA930302-0 VA930503-0 VA930505-0 VA931002-0 VA931005-0 VA931503-0 VA931505-0 VA932505-0
97022-896EA 1333.57 CAD
97022-896 97022-900 97022-902 97022-904 97023-766 97023-768 97023-770 97023-772 97023-774 97023-776 97023-778 75807-354
Venusil AQ C18 HPLC Columns, Bonna-Agela Technologies
Chromatography Columns
The Venusil® AQ C18 column is designed for the separation of polar, medium-polar and non-polar compounds from low to medium pH
  • Single end capped
  • Excellent peak symmetry for basic compounds
  • Large volume injection and maintains very high efficiency
  • 100% water compatible with much better peak shape and retention
Columns offer unmatched inertness, resulting in one of the most base-friendly columns available. These highly efficient columns are excellent at any applicable pH with broad pH range (1.5 - 9.0 ).
With a special surface treatment, Venusil® AQMore Product Information