Ezwaste™ UN/DOT Container with 70 S Cap, Foxx Life Sciences
Supplier: VWR International
75805-548 75805-550 75995-084
75805-548EA 73.3 CAD
75805-548 75805-550 75995-084
Ezwaste™ UN/DOT Container with 70 S Cap, Foxx Life Sciences
Bottles Jerricans
The EZwaste™ UN/DOT System is designed to be an affordable solution for liquid waste disposal.
  • Made from sturdy HDPE
  • Available in multiple sizes
  • Utilizes blow mold technology
The EZwaste™ UN/DOT System empty containers are designed to be used as an affordable solution for liquid waste disposal.
These containers can also be used in other industries, including but not limited to sanitation, chemical, agricultural, and food. All raw materials used in the containers meet FDA Standards for food contact.
These HDPE containers are made to be tough andMore Product Information