Ezwaste™ Solvent Waste System, HD Filter Kit, Safety Vent VersaCap® 83 mm, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0421-OEM 330-0412-OEM 330-0413-OEM 330-0418-OEM 330-0419-OEM
75805-388EA 821.45 CAD
75805-388 75805-480 75805-482 75805-484 75805-486
Ezwaste™ Solvent Waste System, HD Filter Kit, Safety Vent VersaCap® 83 mm, Foxx Life Sciences
Filters Liquid Process Filters
Ezwaste™ Solvent Waste System with the Safety Vent VersaCap® 83 mm is designed to be an affordable solution for HPLC liquid waste disposal.
  • Each system is manufactured to safely collect waste from two HPLC machines
The tube fittings secure waste tubes to the carboy, providing the closed system necessary to prevent spills. Plugs are included for unused ports. A 90° adapter is included for maximum storage flexibility. Finally, the system utilizes a replaceable, sulfur-cured EPDM gasket for high chemical compatibility against aggressive organic solvents.
More Product Information