Forced Air Ovens, SM01, SHEL LAB
Supplier: Sheldon Manufacturing
SLF123 SMO1-2
75802-898EA 7785.61 CAD
75802-898 75802-900
Forced Air Ovens, SM01, SHEL LAB
Ovens Standard Ovens
The SMO1 ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing.
  • Embedded microprocessor control with countdown timer
  • Horizontal forced airflow
  • 1.75" (4.5 cm) access port
  • 24 month limited warranty
They have a quick ramp-up time to 306°C for faster cycle times
This unit is equipped with a stainless steel interior for long life operation and easy cleaning. Its triple wallMore Product Information