Socorex™ Calibrex™ organo/solutae Bottle Top Dispensers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W844090 W844092 W844094 W844102 W844104 W844106 W844096 W844098 W844100 W844108 W844110 W844112 W845016 W845018 W845020 W845022 W845024 W845026 W845028 W845030 W845000 W845002 W845004 W845006 W845008 W845010 W845012 W845014
89496-574CS 2135.67 CAD
89496-574 89496-576 89496-578 89496-580 89496-582 89496-584 10146-734 10146-736 10146-738 10146-740 10146-796 10146-798 75800-442 75800-444 75800-446 75800-484 75800-486 75800-488 75800-490 75800-492 75800-494 75800-496 75800-498 75800-874 75800-876 75800-878 75800-880 75800-882
Socorex™ Calibrex™ organo/solutae Bottle Top Dispensers, DWK Life Sciences
Bottle Top Dispensers
This new generation of Socorex® Bottle Top Dispensers is intended for the safe and reproducible liquid distribution of volumes up to 25, 50, and 100mL
The dispensers are very easy to disassemble and clean. Additionally, they are easy to calibrate in the lab. The calibration mechanism with integrated key is located under the plunger cap. The instructions are engraved for fast and correct setting. Access to this mechanism is protected by a calibration seal sticker.
Dispensers are autoclavable at 121°C fully assembled. The dispensers have a two-year warranty.
The Calibrex™ 525 organoMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable