Corning® Gosselin™ Storage Bottles, Round, HDPE, Corning
Supplier: Corning
LR50A-18 LR250-09 LR500-04 LR100-12 LR1000-08 LR150A-14
75800-328CS 653.46 CAD
75800-328 75800-330 75800-332 75800-334 75800-336 75800-338
Corning® Gosselin™ Storage Bottles, Round, HDPE, Corning
Bottles Plastic Bottles
HDPE, translucent with screw caps.
  • Bottles are supplied capped aseptic or ionised
  • Excellent shock resistance
  • Wide neck, easy to fill
A range of round wide mouth bottles. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.