VWR® TPX Measuring Cylinders, Class B
Supplier: VWR International
346010 346020 346030 346040 346050 346060 346070 346080
75800-218CS 561.32 CAD
75800-218 75800-220 75800-222 75800-224 75800-226 75800-228 75800-230 75800-232
VWR® TPX Measuring Cylinders, Class B
Cylinders Measuring Cylinders
These VWR® Measuring Cylinders are manufactured from translucent polypropylene.
  • The cylinders feature a stable hexagonal base and a pouring spout
  • All cylinders also feature molded graduations for improved accuracy and consistency
When used continuously, the cylinders have a working temperature range of 0 to 121 °C (32 to 248 °F).
Certifications: Products are Class "B" as per DIN 12681 / ISO 6706. Resin meets US FDA 21 CFR requirements.
Order Now
SPECIFICATIONS
Environmentally Preferable