Mobile Phase Reservoir Bottle, Non-Coated, Microsolv
Supplier: MicroSolv Technology Corp
58802-10-X 58802-10-A 58802-05-A 58802-05-X
97057-180EA 1243.29 CAD
97057-180 10804-586 75799-244 75799-246
Mobile Phase Reservoir Bottle, Non-Coated, Microsolv
Bottles HPLC Bottles
These borosilicate glass bottles are non-coated and have a 45 mm standard screw top neck.
Ordering information: Each bottle is supplied with screw cap.
Order Now
SPECIFICATIONS
Environmentally Preferable