Vydac 218TP C18, HPLC Columns
Supplier: Avantor
218TP3410 218TP3215 218TP3415 218TP34 218TP5105 218TP5305 218TP5405 218TP5110 218TP5210 218TP5410 218TP5115 218TP5215 218TP5415 218TP52 218TP53 218TP54 218TP104 218TP1022 5119751 218FSK1210 218TP101510 218TP101522 218TP15202215 218TP152010 218TP152022 218FSK510 218TP51010 218TP510 218GCC1010 218TP1010 218GCC1210 218GD104/N 218GD32/N 218GD34/N 218GD52/N 218GD54/N 218GK104/N 218GK32/N 218GK34/N 218GK52/N 218GK54/N 218TP3405
75798-430EA 2365.37 CAD
75798-430 75798-432 75798-434 75798-436 75798-438 75798-440 75798-442 75798-458 75798-460 75798-462 75798-464 75798-466 75798-468 75798-470 75798-472 75798-474 75798-476 75798-478 75798-480 75798-482 75798-484 75799-122 75799-126 75799-128 75799-130 75799-138 75799-140 75799-142 75799-144 75799-146 75799-148 75799-150 75799-152 75812-900 75812-902 75812-904 75812-906 75812-908 75812-910 75812-912 75813-032 75813-034 75813-036 75813-038 75813-040 75813-042 75813-044 75813-046 75813-048 75813-050 75813-052 75830-782 75831-198 75831-202 75831-398 75831-402 75831-406 75831-410 75831-414 75831-418 75831-422 76398-064 76398-066 76398-076
Vydac 218TP C18, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
First generation polymeric C18 phase with unique selectivity. Recommended for small polypeptides 4-5K MW, enzymatic digest fragments, natural and synthetic peptides, multi-ring compounds.
The large pores ofMore Product Information