Platinum 95%/Gold 5% Dishes, Heraeus
Supplier: HERAEUS MATERIALS TECHNOLOGY NA
SCHA2/1-PTAU5 SCHA2/2-PTAU5 SCHA2/3-PTAU5 SCHA2/4-PTAU5 SCHA2/5-PTAU5 SCHA2/6-PTAU5 SCHA2/7-PTAU5 SCHA2/8-PTAU5 SCHAD1/4 SCHAD1/5 SCHAD1/6 SCHAD1/7 SCHAD1/8
75797-066EA 4713.22 CAD
75797-066 75797-068 75797-070 75797-072 75797-074 75797-076 75797-078 75797-080 75797-112 75797-114 75797-116 75797-118 75797-120
Platinum 95%/Gold 5% Dishes, Heraeus
Trays/Dishes/Pots Evaporating Basins/Dishes
These dishes are made with 95% Platinum and 5% Gold.
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Dishes have become essential for the preparation of samples in analytical laboratories.
The mechanical stability of Platinum is further improved with the addition of 5% Gold. This alloy offers higher strength and lower wettability, so that the sample can be easily removed from the dish.
Ordering information: Pourling Lip available upon request. Lids sold separately. For additional lid details, see Lids for PlatinumMore Product Information