VAI® STER-AHOL® Wipes, Veltek Associates
Supplier: Veltek Associates
VEL6-12X12-S-2320
75795-628CS 900.51 CAD
75795-628
VAI® STER-AHOL® Wipes, Veltek Associates
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
STER-AHOL® Wipes may be used as a sanitizer in classified manufacturing and testing environments.
  • 100% continuous knit polyester filament is specifically designed to be clean and absorbent
  • Gamma irradiated at 10⁻⁶ SAL
  • Completetly traceable from start to finish
  • Individually bagged with two liner bags
STER-AHOL® Wipes are saturated with denatured ethanol formulated to 70%, with a small percentage of methyl alcohol and 30% USP water for injection. These wipes can be used in both aseptic and non-aseptic wipe downs on any critical surface that requires cleaning.