Lysing Matrix B, MP Biomedicals

Supplier: MP Biomedicals
116911050 116911100 116911500 116971050 116971100 116971025 116981001 116981010 116911100 116911500 116540425 116540429
75784-614EA 253.7 CAD
75784-614 75784-616 75784-618 CA76045-728 CA76045-730 CA76045-732 CA76045-770 CA76045-772 CA76046-064 CA76046-066 CA76046-224 CA76046-226
Lysing Matrix B, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores

  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria

Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then preparedMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Tube compatibility 2 mL 2 ml 2 ml 2 ml 2 ml 2 ml 2 ml 2 ml 2 ml
Specific gravity 2.5 2.5 2.5 2.5 2.5 2.5 2.5 2.5 2.5
Storage and handling Store at Room Temperature (15 to 30° C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C) Store at Room Temperature (15...30 °C)
Sample type Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores Bacteria (Gram +/-) and Spores
Molecule enriched DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules DNA, RNA, Proteins, Other small molecules
Materials required (not supplied) FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge, Pipettmen, Purification Reagents
Material of construction 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres 0.1mm silica spheres
Key applications Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria Lysis of bacteria
Instrument Compatibility FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-96 FastPrep-96 FastPrep-24; FastPrep 120; FastPrep-96 FastPrep-24, FastPrep-96, SuperFastPrep-2, FastPrep-24 5G
Adapter compatibility QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96) QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96) QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96) TallPrep (24×4.5 ml), TallFlex (48×4.5 ml), ConeFlex Legacy (allows all FP24 adapters on FP96) TallPrep (24×4.5 ml), TallFlex (48×4.5 ml), ConeFlex Legacy (allows all FP24 adapters on FP96) 96 Well Adapter 96 Well Adapter QuickPrep (24×2 ml), HiPrep (48×2 ml), CoolPrep (24×2 ml), QuickFlex (96×2 ml), ConeFlex Legacy (allows all FP24 adapters on FP96) All FastPrep Adapters
Application areas Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification Nucleic Acid Isolation & Purification

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR