Corning® CoolRack® Modules, Corning
Supplier: Corning
432046 432047 432052 432053 432054 432055 432056 432057 432058 432059 432060 432061 432062 432063 432064 432065 432066 432067 432068 432069 432070 432071 432072 432048 432049 432050 432051 432034 432035 432036 432037 432038 432039 432040 432041 432042 432043 432044 432045 432139
75779-726CS 552.59 CAD
75779-726 75779-728 75779-730 75779-732 75779-734 75779-736 75779-738 75779-740 75779-742 75779-744 75779-746 75779-748 75779-750 75779-752 75779-754 75779-756 75779-758 75779-760 75779-762 75779-764 75779-766 75779-768 75779-770 75779-832 75779-834 75779-836 75779-838 75779-878 75779-880 75779-882 75779-884 75779-886 75779-888 75779-890 75779-892 75779-894 75779-896 75779-898 75779-900 CA76332-748
Corning® CoolRack® Modules, Corning
Racks Cooling Racks
These thermoconductive modules provide reproducible and standardised temperature control to biological samples while on the bench.
  • Place CoolRack® or CoolSink™ modules on ice, dry ice, liquid nitrogen, water baths, heat blocks (–196 to 100 °C), or other vessels and they rapidly adapt to source temperature
Cool, (snap) freeze, or thaw samples consistently and reproducibly without directly inserting them into a temperature source to reduce variability and contamination. CoolRack® tube holders and CoolSink™ plate holders accommodate a wide variety ofMore Product Information