Corning® CoolCell® Containers, Corning
Supplier: Corning
432000 432001 432002 432003 432004 432005 432006 432007 432008 432009 432010 432011 432138
75779-712CS 335.98 CAD
75779-712 75779-714 75779-716 75779-718 75779-720 75779-722 75779-724 75779-814 75779-816 75779-818 75779-820 75779-822 76180-494
Corning® CoolCell® Containers, Corning
Boxes Cryoboxes
CoolCell® alcohol-free cell freezing containers ensure standardized controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.
  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 ˚C freezer
  • Quick re-use time, ready to use again after five minutes
Proven for use with a varietyMore Product Information