Recessed Single Taper Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Supplier: Restek
20984 20981 20984-214
RK20984PK 207.51 CAD
RK20984 RK20981 75778-398
Recessed Single Taper Inlet Liner for Agilent GCs Equipped with Split/Splitless Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Order Now
SPECIFICATIONS
Environmentally Preferable