Guardian™ Airflow Monitor Kits, Labconco®
Supplier: Labconco
9743203 9743205 9743207 9743209 9743208 9743210 3908800 3908801 9743201 3908801 9743201 9743207
46620-470EA 5506.3 CAD
46620-470 74920-162 82001-232 82007-280 82007-282 82007-284 82007-286 82010-706 82010-708 14222-630 470203-190 470203-272 470203-276
Guardian™ Airflow Monitor Kits, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Guardian™ airflow monitors continuously check the face velocity of laboratory hoods and enclosures and alert the operator to airflows that drop outside preset levels with audible and visual alarms. The audible alarm can be muted at the touch of a switch.
The Guardian™ digital monitor displays actual face velocity on an LCD screen and includes a programmable switch to open the hood damper
The right fixture panel is factory-prepared for an airflow monitor on the following nonexplosion-proof laboratory hoods: Protector Premier™, Protector XStream®, Protector XL™, Protector Stainless Steel,More Product Information