Nalgene® Square Media Bottles, PET, Sterile, Graduated, Thermo Scientific
Supplier: Thermo Fisher Scientific
342044-0650 342040-0650 342040-1000 342044-1000 342040-0125 342044-0125 342040-0250
16010-100CS 275.6 CAD
16010-100 16010-102 16010-104 16010-106 73520-442 73520-444 73521-216
Nalgene® Square Media Bottles, PET, Sterile, Graduated, Thermo Scientific
Bottles Cell Culture Bottles
Bottles are lightweight, shatter-resistant, and provide excellent gas barrier properties for storage and shipping of liquid media, buffers, and sera.
  • Noncytotoxic
Sterile and nonpyrogenic to save time and prevent costly cleaning preparation. Bottles feature molded-in graduations and molded-in neck ring for use with tamper-evident shrink sleeves. Suitable for air shipment and tested to ensure leakproof performance.
Certifications: Bottles meet USP Class VI, USP 661, and European Pharmacopeia for Abnormal Toxicity standards.
Ordering information: More Product Information