Nalgene® High-Density Polyethylene Economy Bottles, Bulk Pack, Wide Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
332189-0004 332189-0008 332189-0016 332189-0032 332189-0002
16060-008CS 935.36 CAD
16060-008 16060-010 16060-012 16060-014 73520-258
Nalgene® High-Density Polyethylene Economy Bottles, Bulk Pack, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
  • Priced below low-quality plastic or glass bottle brands
  • Economical, quality choice for lighter-duty lab use
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent high-density polyethylene. Like most Nalgene bottles, the Economy Bottles are guaranteed leakproof and are made in our ISO-certified USAMore Product Information