Microliter™ Micro Syringe Pipette, Hamilton Company
Supplier: Hamilton
84250
72310-316EA 309.36 CAD
72310-316
Microliter™ Micro Syringe Pipette, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Micro Syringe Pipettes have a special 3/4" electrotapered needle for easy attachment of PTFE tips. The sample only wets the disposable PTFE tips, eliminating sample cross-contamination. The spring-loaded plunger and adjustable stops ensure excellent reproducibility. An adjustable stop allows the transfer of samples from 0.2µL to 03µL. Syringes in this category are 700 series Microliter™ syringes. They have a stainless steel plunger that is hand fitted to the glass barrel and areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable