Tote Box, Polypropylene, Dynalon
Supplier: Dynalon
410545 410535
71070-044CS 575.99 CAD
71070-044 470173-918
Tote Box, Polypropylene, Dynalon
Boxes Transport Boxes
Sterilizing Tote Boxes, molded of natural Polypropylene is suitable for use with food
These tote boxes are chemically resistant to acids, alkalies, oils and detergents and will not rust or corrode. Can be nested when stored.
Order Now
SPECIFICATIONS
Environmentally Preferable